CAS RN 21373-30-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ 6-Hydroxy-DL-DOPA
Allosteric inhibitor of RAD52; also APE1 inhibitor
2
3