missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 6-Hydroxy-DL-DOPA

Description
6-Hydroxy-DL-DOPA is an allosteric inhibitor of RAD52; inhibits RAD52 binding to single strand DNA binding domains (IC50 = 1.1 μM). Selectively inhibits proliferation of BRCA-deficient cancer cells in vitro. Also inhibits APE1.
Specifications
Specifications
| Chemical Name or Material | 2,5-Dihydroxy-DL-tyrosine |
| CAS | 21373-30-8 |
| Quantity | 10 mg |
| Target | DNA, RNA and Protein Synthesis Inhibitors |
| Molecular Formula | C9H11NO5 |
| Formula Weight | 213.19 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?