Learn More
Tocris Bioscience™ 6-Hydroxy-DL-DOPA
Allosteric inhibitor of RAD52; also APE1 inhibitor
Brand: Tocris Bioscience™ 5740/10
This item is not returnable.
View return policy
Description
6-Hydroxy-DL-DOPA is an allosteric inhibitor of RAD52; inhibits RAD52 binding to single strand DNA binding domains (IC50 = 1.1 μM). Selectively inhibits proliferation of BRCA-deficient cancer cells in vitro. Also inhibits APE1.Specifications
| 2,5-Dihydroxy-DL-tyrosine | |
| 21373-30-8 | |
| 10 mg |