Learn More
Tocris Bioscience™ 6-Hydroxy-DL-DOPA
GREENER_CHOICE
Allosteric inhibitor of RAD52; also APE1 inhibitor
Brand:  Tocris Bioscience™ 5740/10
Encompass
Product Code. 15757086
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
6-Hydroxy-DL-DOPA is an allosteric inhibitor of RAD52; inhibits RAD52 binding to single strand DNA binding domains (IC50 = 1.1 μM). Selectively inhibits proliferation of BRCA-deficient cancer cells in vitro. Also inhibits APE1.
Specifications
Specifications
2,5-Dihydroxy-DL-tyrosine
21373-30-8