accessibility menu, dialog, popup

UserName

CAS RN 99519-84-3

NH2ONNNClClOClH2N

CAS RN 99519-84-3

IUPAC Name: 5-amino-1-{[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl}-1H-1,2,3-triazole-4-carboxamide
Synonyms: carboxyamidotriazole
Molecular Weight (g/mol): 424.67
Molecular Formula: C17H12Cl3N5O2
InChi Key: WNRZHQBJSXRYJK-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)N(CC2=CC(Cl)=C(C(=O)C3=CC=C(Cl)C=C3)C(Cl)=C2)N=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results