missing translation for 'onlineSavingsMsg'
Learn More

Carboxyamidotriazole, TRC

Product Code. 30361853
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361853 5 mg 5mg
30353406 50 mg 50mg
30355502 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361853 Supplier Toronto Research Chemicals Supplier No. TRCC1777755MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99519-84-3
Molecular Formula C17 H12 Cl3 N5 O2
Molecular Weight (g/mol) 424.67
Synonym 1H-1,2,3-Triazole-4-carboxamide, 5-amino-1-[[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl]-, 5-Amino-1-[[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl]-1H-1,2,3-triazole-4-carboxamide, CAI, Carboxyamidotriazole, L 651582, NSC 609974
IUPAC Name 5-amino-1-[[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl]triazole-4-carboxamide
SMILES NC(=O)c1nnn(Cc2cc(Cl)c(C(=O)c3ccc(Cl)cc3)c(Cl)c2)c1N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H12Cl3N5O2/c18-10-3-1-9(2-4-10)15(26)13-11(19)5-8(6-12(13)20)7-25-16(21)14(17(22)27)23-24-25/h1-6H,7,21H2,(H2,22,27)
Formula Weight 423.0057
Chemical Name or Material Carboxyamidotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.