missing translation for 'onlineSavingsMsg'
Learn More

Carboxyamidotriazole, MedChemExpress

Product Code. 16411599
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411599 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411599 Supplier Medchem Express Supplier No. HY16126
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Carboxyamidotriazole (L-651582) is a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways. Carboxyamidotriazole shows anti-tumor, anti-inflammatory and antiangiogenic effects.

Carboxyamidotriazole (L-651582) is a cytostatic inhibitor of nonvoltage-operated calcium channels and calcium channel-mediated signaling pathways. Carboxyamidotriazole shows anti-tumor, anti-inflammatory and antiangiogenic effects.

Specifications

Chemical Name or Material Carboxyamidotriazole
Physical Form Solid
CAS 99519-84-3
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C17H12Cl3N5O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym L-651582 CAI
Solubility Information DMSO : 100 mg/mL (235.48 mM; Need ultrasonic)
SMILES O=C(C1=C(N)N(CC2=CC(Cl)=C(C(C3=CC=C(Cl)C=C3)=O)C(Cl)=C2)N=N1)N
Molecular Weight (g/mol) 424.67
Formula Weight 424.67
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.