accessibility menu, dialog, popup

UserName

CAS RN 71048-99-2

NaCH3NHOCH3HNONH2PH3CHOOOO

CAS RN 71048-99-2

IUPAC Name: sodium 2-(2-{2-amino-4-[hydroxy(methyl)phosphoryl]butanamido}propanamido)propanoate
Synonyms: sodium 2-(2-{2-amino-4-[hydroxy(methyl)phosphoryl]butanamido}propanamido)propanoate
Molecular Weight (g/mol): 345.27
Molecular Formula: C11H21N3NaO6P
InChi Key: RTWIRLHWLMNVCC-UHFFFAOYNA-M
SMILES: [Na+].CC(NC(=O)C(C)NC(=O)C(N)CCP(C)(O)=O)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
Color 
  • (1)
  • (2)
Industry Type 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)