missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Bialaphos ReadyMade™ Solution

Product Code. 30044044
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044044 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044044 Supplier TOKU-E™ Supplier No. B10010x1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bialaphos ReadyMade™ Solution is provided as a sterile-filtered solution of Bialaphos Sodium in water.

  • CAS Number: 71048-99-2
  • Molecular Formula: C11H21N3O6S2P · Na
  • Molecular Weight: 345.27 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces griseochromogenes
  • Spectrum: Bialaphos is a broad-spectrum herbicide that also has activity against Gram-positive and Gram-negative bacteria, and certain fungi
  • Mechanism of Action: Bialaphos acts as a pro-herbicide by separating into individual subunits of alanylalanine and phosphinothricin. Phosphinothricin is toxic to cells as it triggers the accumulation of ammonia, thus inhibiting photosynthesis and glutamine synthesis

Specifications

Chemical Name or Material Bialaphos sodium
CAS 71048-99-2
Assay Percent Range >80.0%
Synonym Bialaphos sodium salt solution
Quantity 10 x 1 mL
Solubility Freely soluble in water
Shipping Condition Ice Pack
Detection Method HPLC
Product Line ReadyMade™ Solutions
Target Photosynthesis and glutamine synthesis
Molecular Formula C11H21N3O6P ∙ Na
Formula Weight 345.27 amu
Physical Form Liquid
Color Clear
Product Type Antibiotic
Purity >80%
Molecular Weight (g/mol) 345.27 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.