Learn More
Bialaphos sodium, 85%, Thermo Scientific™
Bialaphos sodium (bilanafos), CAS # 35597-43-4, is a tripeptide antibiotic used to confirm gene transfer in plants through use of the bar selection gene.
272.00 EUR - 1076.00 EUR
Chemical Identifiers
CAS 71048-99-2
Molecular Formula C11H21N3NaO6P
Molecular Weight (g/mol) 345.268
MDL Number MFCD01674432
InChI Key RTWIRLHWLMNVCC-WQYNNSOESA-M
PubChem CID 118988468
IUPAC Name sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
SMILES CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428309
SDS View Documents
Thermo Scientific Alfa Aesar
J66802.MC
100mg
272.00 EUR
100mg
View alternative products
Save Up to 
»
15438309
SDS View Documents
Thermo Scientific Alfa Aesar
J66802.ME
500mg
1076.00 EUR
500mg
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
• Bialaphos is a bacterial metabolite and an antimicrobial
Application
• Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
• Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
Specifications
Chemical Identifiers
71048-99-2
345.268
RTWIRLHWLMNVCC-WQYNNSOESA-M
sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
C11H21N3NaO6P
MFCD01674432
118988468
CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]
Specifications
71048-99-2
>110°C (230°F)
MFCD01674432
793°C
RTWIRLHWLMNVCC-WQYNNSOESA-M
sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
118988468
85%
Orange
C11H21N3NaO6P
UN2811
Soluble in water
CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]
345.268
345.26
Bialaphos sodium
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : AY6827000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold