accessibility menu, dialog, popup

UserName

OONOONOO

CAS RN 64987-85-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexane-1-carboxylate
Synonyms: SMCC
Molecular Weight (g/mol): 334.33
Molecular Formula: C16H18N2O6
InChi Key: JJAHTWIKCUJRDK-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)C1CCC(CN2C(=O)C=CC2=O)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results