Learn More
N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 98+%
Product Code. 15441416
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441416 100 mg 100mg
15431416 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15441416 Supplier Thermo Scientific Alfa Aesar Supplier No. H54627.MC
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 64987-85-5 | C16H18N2O6 | 334.328 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64987-85-5
Molecular Formula C16H18N2O6
Molecular Weight (g/mol) 334.328
MDL Number MFCD00009634
InChI Key JJAHTWIKCUJRDK-UHFFFAOYSA-N
Synonym smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
PubChem CID 125175
ChEBI CHEBI:63174
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
SMILES C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O
Specifications
Melting Point 180°C to 182°C
Quantity 100 mg
Beilstein 1555271
Sensitivity Light and moisture sensitive
Formula Weight 334.33
Percent Purity ≥98%
Chemical Name or Material N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate