Learn More
N-Succinimidyl 4-(Maleimidomethyl)cyclohexane-1-carboxylate, TRC
Product Code. 30412351
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412351 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30412351 Supplier Toronto Research Chemicals Supplier No. TRCS6904001G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Succinimidyl 4-(Maleimidomethyl)cyclohexane-1-carboxylate
CAS 64987-85-5
Molecular Formula C16 H18 N2 O6
Quantity 1 g
InChI Formula InChI=1S/C16H18N2O6/c19-12-5-6-13(20)17(12)9-10-1-3-11(4-2-10)16(23)24-18-14(21)7-8-15(18)22/h5-6,10-11H,1-4,7-9H2
SMILES O=C(ON1C(=O)CCC1=O)C2CCC(CN3C(=O)C=CC3=O)CC2
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
Molecular Weight (g/mol) 334.32
Formula Weight 334.1165