Learn More
N-Succinimidyl 4-(Maleimidomethyl)cyclohexane-1-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | N-Succinimidyl 4-(Maleimidomethyl)cyclohexane-1-carboxylate |
| CAS | 64987-85-5 |
| Molecular Formula | C16 H18 N2 O6 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C16H18N2O6/c19-12-5-6-13(20)17(12)9-10-1-3-11(4-2-10)16(23)24-18-14(21)7-8-15(18)22/h5-6,10-11H,1-4,7-9H2 |
| SMILES | O=C(ON1C(=O)CCC1=O)C2CCC(CN3C(=O)C=CC3=O)CC2 |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate |
| Molecular Weight (g/mol) | 334.32 |
| Formula Weight | 334.1165 |