CAS RN 63-91-2
Thermo Scientific Chemicals L-Phenylalanine, 99%
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD00064227 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 SMILES: NC(CC1=CC=CC=C1)C(O)=O
Thermo Scientific Chemicals L-Phenylalanine, 98.5+%
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD00064227 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 SMILES: NC(CC1=CC=CC=C1)C(O)=O
Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD00064227 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 SMILES: NC(CC1=CC=CC=C1)C(O)=O
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 SMILES: NC(CC1=CC=CC=C1)C(O)=O
L-phenylalanine, 99.85%, MP Biomedicals™
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 IUPAC Name: 2-amino-3-phenylpropanoic acid SMILES: NC(CC1=CC=CC=C1)C(O)=O
L-Phenylalanine, >99%, MP Biomedicals
CAS: 63-91-2 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 InChI Key: COLNVLDHVKWLRT-UHFFFAOYNA-N Synonym: l-phenylalanine,phenylalanine,s-2-amino-3-phenylpropanoic acid,3-phenyl-l-alanine,s-phenylalanine,3-phenylalanine,2s-2-amino-3-phenylpropanoic acid,s-2-amino-3-phenylpropionic acid,beta-phenyl-l-alanine,h-phe-oh PubChem CID: 6140 ChEBI: CHEBI:17295 IUPAC Name: 2-amino-3-phenylpropanoic acid SMILES: NC(CC1=CC=CC=C1)C(O)=O
l-Phenylalanine, Millipore™
CAS: 63-91-2
L-Phenylalanine, TRC
CAS: 63-91-2 Molecular Formula: C9 H11 N O2 Molecular Weight (g/mol): 165.19 Synonym: L-Phenylalanine,Alanine, phenyl-, L- (4CI,7CI,8CI),(-)-Phenylalanine,(2S)-2-Amino-3-phenylpropanoic acid,(2s)-2-Amino-3-phenylpropanoic acid,(S)-(-)-Phenylalanine,(S)-2-Amino-3-phenylpropanoic acid,(S)-2-Amino-3-phenylpropionic acid,(S)-Phenylalanine,(S)-α-Amino-β-phenylpropionic acid,(S)-α-Aminobenzenepropanoic acid,(S)-α-Aminohydrocinnamic acid,3-Phenyl-L-alanine,3-Phenylalanine,Antibiotic FN 1636,Benzenepropanoic acid, α-amino-, (S)-,L-(-)-Phenylalanine,L-Alanine, 3-phenyl-,NSC 79477,Phenyl-α-alanine,Phenylalanine,β-Phenyl-L-alanine,β-Phenyl-α-alanine,Aspartame Imp. C (EP) IUPAC Name: (2S)-2-amino-3-phenylpropanoic acid SMILES: N[C@@H](Cc1ccccc1)C(=O)O
Phenylalanine (L-Phenylalanine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(S)-(-)-Phenylalanine, Sigma-Aldrich™
CAS: 63-91-2 Grade: Research Molecular Weight (g/mol): 165.19
L-Phenylalanine, MedChemExpress
MedChemExpress L-Phenylalanine ((S)-2-Amino-3-phenylpropionic acid) is an essential amino acid isolated from Escherichia coli.
Phenylalanine (L-Phenylalanine), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.