missing translation for 'onlineSavingsMsg'
Learn More

l-Phenylalanine, Millipore™

Product Code. 10795761
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10795761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795761 Supplier Millipore Supplier No. 1072560100
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2

Specifications

Melting Point 275°C to 283°C (decomposition)
MDL Number MFCD00064227
Synonym L-Phenylalanine, Phe, 2-Amino-3-phenylpropionic acid, L-Phenylalanine
Density 1.34 g/cm3 at 20°C
Molecular Weight (g/mol) 165.19
Quantity 100 g
Grade Research
Chemical Name or Material L-Phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.