accessibility menu, dialog, popup

UserName

CAS RN 39809-25-1

H2NNONNOHOHNH

CAS RN 39809-25-1

IUPAC Name: 2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6,9-dihydro-3H-purin-6-one
Synonyms: penciclovir penciclovir, monosodium salt sb-ch brand of penciclovir monosodium salt penciclovir vectavir novartis brand of penciclovir denavir visfarm brand of penciclovir penciclovir, monosodium salt, monohydrate penciclovir visfarm
Molecular Weight (g/mol): 253.26
Molecular Formula: C10H15N5O3
InChi Key: JNTOCHDNEULJHD-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(N1)N(CCC(CO)CO)C=N2
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)