missing translation for 'onlineSavingsMsg'
Learn More

Penciclovir, MedChemExpress

Product Code. 16497296
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497296 100 mg 100mg
16493347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497296 Supplier Medchem Express Supplier No. HY17424
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Penciclovir (VSA 671) is a potent and selective anti-herpesvirus agent with EC50 values of 0.5, 0.8 μg/ml for HSV-1 (HFEM), HSV-2 (MS), respectively. Penciclovir shows anti-herpesvirus activity with no-toxic. Penciclovir preventes mortality in mouse.

Penciclovir (VSA 671) is a potent and selective anti-herpesvirus agent with EC50 values of 0.5, 0.8 μg/ml for HSV-1 (HFEM), HSV-2 (MS), respectively. Penciclovir shows anti-herpesvirus activity with no-toxic. Penciclovir preventes mortality in mouse.

Specifications

Chemical Name or Material Penciclovir
CAS 39809-25-1
Color White
Purity Grade Notes Research
Molecular Formula C10H15N5O3
Quantity 100 mg
Synonym BRL 39123 VSA 671
Solubility Information DMSO : 25 mg/mL (98.71 mM; Need ultrasonic)
SMILES O=C1NC(N)=NC2=C1N=CN2CCC(CO)CO
Molecular Weight (g/mol) 253.26
Formula Weight 253.26
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.