missing translation for 'onlineSavingsMsg'
Learn More

Penciclovir, TRC

Product Code. 30138463
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138463 250 mg 250mg
30133312 500 mg 500mg
30011370 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138463 Supplier Toronto Research Chemicals Supplier No. TRCP221500250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39809-25-1
Molecular Formula C10 H15 N5 O3
Molecular Weight (g/mol) 253.26
Synonym 2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one, Denavir, Penciclovir, VSA 671;
IUPAC Name 2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-3H-purin-6-one
SMILES NC1=NC(=O)c2ncn(CCC(CO)CO)c2N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H15N5O3/c11-10-13-8-7(9(18)14-10)12-5-15(8)2-1-6(3-16)4-17/h5-6,16-17H,1-4H2,(H3,11,13,14,18)
Formula Weight 253.1175
Percent Purity >95%
Chemical Name or Material Penciclovir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.