CAS RN 26016-99-9
IUPAC Name:
disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
Synonyms:
disodium fosfomycin(2-)
Molecular Weight (g/mol):
182.02
Molecular Formula:
C3H5Na2O4P
InChi Key:
QZIQJIKUVJMTDG-JSTPYPERSA-L
SMILES:
[Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O
1
–
3
of
3
results
2
Phosphonomycin disodium salt, 100.4%, MP Biomedicals™
CAS: 26016-99-9 Molecular Formula: C3H5Na2O4P Molecular Weight (g/mol): 182.02 MDL Number: MFCD00056853 InChI Key: QZIQJIKUVJMTDG-JSTPYPERSA-L Synonym: (2R-cis)-(3-Methyloxiranyl) phosphonic acid disodium salt,Fosfomycin disodium PubChem CID: 16397688 IUPAC Name: disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate SMILES: [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O
3
Fosfomycin Disodium Salt, TRC
CAS: 26016-99-9 Molecular Formula: C3 H5 O4 P . 2 Na Molecular Weight (g/mol): 182.02 Synonym: Fosfomycin sodium,Disodium (2R,3S)-(3-methyloxiran-2-yl)phosphonate IUPAC Name: disodium;[(2R,3S)-3-methyloxiran-2-yl]-dioxido-oxo-λ^{5}-phosphane SMILES: [Na+].[Na+].C[C@@H]1O[C@@H]1P(=O)([O-])[O-]