missing translation for 'onlineSavingsMsg'
Learn More

Phosphomycin Disodium Salt, MP Biomedicals

Product Code. 11497092 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497092 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497092 Supplier MP Biomedicals Supplier No. 0215187601
  • ONLINE EXCLUSIVE PRICE
    52.95 EUR
    50.40 EUR / 1g
    Save 2.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 26016-99-9
Quantity 1 g
Molecular Formula C3H5Na2O4P
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
Molecular Weight (g/mol) 182.02
PubChem CID 16397688
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.