missing translation for 'onlineSavingsMsg'
Learn More

Fosfomycin Disodium Salt, TRC

Product Code. 30007277
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007277 1 g 1g
30011296 10 g 10g
17964255 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007277 Supplier Toronto Research Chemicals Supplier No. TRCF7275001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3 H5 O4 P . 2 Na
Molecular Weight (g/mol) 182.02
Synonym Fosfomycin sodium, Disodium (2R,3S)-(3-methyloxiran-2-yl)phosphonate
IUPAC Name disodium;[(2R,3S)-3-methyloxiran-2-yl]-dioxido-oxo-λ^{5}-phosphane
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P(=O)([O-])[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H7O4P.2Na/c1-2-3(7-2)8(4,5)6;;/h2-3H,1H3,(H2,4,5,6);;/q;2*+1/p-2/t2-,3+;;/m0../s1
Formula Weight 181.9721
Percent Purity >95%
Chemical Name or Material Fosfomycin Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.