missing translation for 'onlineSavingsMsg'
Learn More

Phosphonomycin disodium salt, 100.4%, MP Biomedicals™

Product Code. 11417102 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417102 10 g 10g
11407102 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417102 Supplier MP Biomedicals Supplier No. 0215187610
  • ONLINE EXCLUSIVE PRICE
    201.00 EUR
    191.10 EUR / 10g
    Save 9.90 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3H5Na2O4P
Molecular Weight (g/mol) 182.02
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
Synonym (2R-cis)-(3-Methyloxiranyl) phosphonic acid disodium salt, Fosfomycin disodium
PubChem CID 16397688
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

Specifications

Type Phosphonomycin Disodium Salt
Physical Form Powder
Color White
Quantity 10 g
Assay Percent Range 1.004
Formula Weight 182
Chemical Name or Material Phosphonomycin disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.