accessibility menu, dialog, popup

UserName

CAS RN 2113-55-5

IUPAC Name: [1,1'-biphenyl]-3-amine hydrochloride
Synonyms: [1,1'-biphenyl]-3-amine hydrochloride
Molecular Weight (g/mol): 205.69
Molecular Formula: C12H12ClN
InChi Key: FFNBBUJVAUTTPK-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=CC(=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)