Learn More
3-Aminobiphenyl hydrochloride, Thermo Scientific™
Product Code. 15417917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417917 5 g 5g
15407917 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15417917 Supplier Thermo Scientific Alfa Aesar Supplier No. H51027.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2113-55-5
Molecular Formula C12H12ClN
Molecular Weight (g/mol) 205.685
MDL Number MFCD00068358
InChI Key FFNBBUJVAUTTPK-UHFFFAOYSA-N
Synonym 1,1'-biphenyl-3-amine hydrochloride, 3-aminobiphenyl hydrochloride, 3-phenylaniline hydrochloride, 1,1'-biphenyl-3-amine, hydrochloride, biphenyl-3-amine hcl, 3-aminobiphenyl, hcl, m-xenylamine hydrochloride, 3-biphenylamine hydrochloride, biphenyl-3-amine hydrochloride, biphenyl-3-ylamine hydrochloride
PubChem CID 12219023
IUPAC Name 3-phenylaniline;hydrochloride
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)N.Cl
Specifications
Odor Odorless
Quantity 5 g
Formula Weight 205.69
Chemical Name or Material 3-Aminobiphenyl hydrochloride