Learn More
3-Aminobiphenyl Hydrochloride, TRC
Product Code. 30327315
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327315 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327315 Supplier Toronto Research Chemicals Supplier No. TRCA6017742G
 Restricted Item
Estimated Shipment: 05-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Aminobiphenyl Hydrochloride
CAS 2113-55-5
Molecular Formula C12 H11 N . HCl
Quantity 2 g
Synonym 3-Aminobiphenyl HCl
InChI Formula InChI=1S/C12H11N.ClH/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10;/h1-9H,13H2;1H
SMILES Cl.NC1=CC=CC(=C1)C=2C=CC=CC2
IUPAC Name 3-phenylaniline;hydrochloride
Molecular Weight (g/mol) 205.68338
Formula Weight 205.0658