Learn More
3-Aminobiphenyl Hydrochloride, TRC
Product Code. 30319827
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319827 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30319827 Supplier Toronto Research Chemicals Supplier No. TRCA601774100MG
 Restricted Item
Estimated Shipment: 05-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Aminobiphenyl Hydrochloride
CAS 2113-55-5
Molecular Formula C12 H11 N . HCl
Quantity 100 mg
Synonym 3-Aminobiphenyl HCl
InChI Formula InChI=1S/C12H11N.ClH/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10;/h1-9H,13H2;1H
SMILES Cl.NC1=CC=CC(=C1)C=2C=CC=CC2
IUPAC Name 3-phenylaniline;hydrochloride
Molecular Weight (g/mol) 205.68338
Formula Weight 205.0658