accessibility menu, dialog, popup

UserName

CAS RN 2035509-96-5

H3COONNNCH3NHNNNONCH3SOOCH3

CAS RN 2035509-96-5

IUPAC Name: 5-({2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2-methanesulfonyl-9-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 5-({2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2-methanesulfonyl-9-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 648.78
Molecular Formula: C32H40N8O5S
InChi Key: HTFNVAVTYILUCF-UHFFFAOYSA-N
SMILES: CCOC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C3=N2)S(C)(=O)=O)C=CC(=C1)C(=O)N1CCC(CC1)N1CCN(C)CC1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results