Learn More
AX-15836, MedChemExpress
Product Code. 16473608
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473608 10 mg 10mg
16472786 25 mg 25mg
16446337 5 mg 5mg
16440066 2 mg 2mg
16410007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16473608 Supplier Medchem Express Supplier No. HY101846
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
MedChemExpress AX-15836 is a potent and selective ERK5 inhibitor with an IC50 of 8 nM.
AX-15836 is a potent and selective ERK5 inhibitor with an IC50 of 8 nM.
Specifications
Chemical Name or Material AX-15836
CAS 2035509-96-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H40N8O5S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (154.14 mM; Need ultrasonic)
SMILES O=C1N(C)C2=CN=C(NC3=CC=C(C(N4CCC(N5CCN(C)CC5)CC4)=O)C=C3OCC)N=C2N(S(=O)(C)=O)C6=CC=CC=C16
Molecular Weight (g/mol) 648.78
Formula Weight 648.78
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease