CAS RN 1781882-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Pyridostatin hydrochloride, MedChemExpress
MedChemExpress Pyridostatin (RR82) hydrochloride is a G-quadruplex DNA stabilizing agent (Kd=490 nM). Pyridostatin hydrochloride promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. Pyridostatin hydrochloride targets the proto-oncogene Src. Pyridostatin hydrochloride reduced SRC protein levels and SRC-dependent cellular motility in human breast cancer cells.
2
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest
3
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest