Learn More
Tocris Bioscience™ Pyridostatin pentahydrochloride
GREENER_CHOICE
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest
Brand:  Tocris Bioscience™ 4763/10
Encompass
Product Code. 15736575
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).
Specifications
Specifications
95%
1781882-65-2
10 mg
4-(2-Aminoethoxy)N2,N6-bis[(4-(2-aminoethoxy)-2-quinolinyl]-2,6-pyridinecarboxamide pentahydrochloride
Desiccate at RT