accessibility menu, dialog, popup

UserName

CAS RN 150683-30-0

H3CONHH3CON(R)HOCl

CAS RN 150683-30-0

IUPAC Name: N-{4-[(5R)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1H-1-benzazepine-1-carbonyl]-3-methylphenyl}-2-methylbenzamide
Synonyms: tolvaptan
Molecular Weight (g/mol): 448.95
Molecular Formula: C26H25ClN2O3
InChi Key: GYHCTFXIZSNGJT-XMMPIXPASA-N
SMILES: CC1=CC=CC=C1C(=O)NC1=CC=C(C(=O)N2CCC[C@@H](O)C3=CC(Cl)=CC=C23)C(C)=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results