Learn More
Tolvaptan, TRC
Product Code. 30011577
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011577 5 mg 5mg
30010425 25 mg 25mg
30009375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011577 Supplier Toronto Research Chemicals Supplier No. TRCT5366505MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 150683-30-0
Molecular Formula C26 H25 Cl N2 O3
Molecular Weight (g/mol) 448.94
Synonym N-[4-[(7-Chloro-2,3,4,5-tetrahydro-5-hydroxy-1H-1-benzazepin-1-yl)carbonyl]-3-methylphenyl]-2-methyl-benzamide, 1H-1-Benzazepine, benzamide deriv., 1-[4-(2-Methylbenzoylamino)-2-methylbenzoyl]-7-chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-ol, OPC 41061, Samsca, Tolvaptan
IUPAC Name N-[4-(7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl)-3-methylphenyl]-2-methylbenzamide
SMILES Cc1ccccc1C(=O)Nc2ccc(C(=O)N3CCCC(O)c4cc(Cl)ccc34)c(C)c2
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C26H25ClN2O3/c1-16-6-3-4-7-20(16)25(31)28-19-10-11-21(17(2)14-19)26(32)29-13-5-8-24(30)22-15-18(27)9-12-23(22)29/h3-4,6-7,9-12,14-15,24,30H,5,8,13H2,1-2H3,(H,28,31)
Formula Weight 448.1554
Chemical Name or Material Tolvaptan