missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tolvaptan

Product Code. 17291146 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17291146 250 mg 250mg
17281146 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17291146 Supplier Thermo Scientific Chemicals Supplier No. 465352500
  • ONLINE EXCLUSIVE PRICE
    166.00 EUR
    149.65 EUR / 250mg
    Save 16.35 EUR
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 150683-30-0 | C26H25ClN2O3, C26H25ClN2O3 | 448.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150683-30-0
Molecular Formula C26H25ClN2O3

Specifications

Quantity 250 mg
Melting Point 225.9°C
Formula Weight 448.95
InChI Key GYHCTFXIZSNGJT-XMMPIXPASA-N
SMILES CC1=CC=CC=C1C(=O)NC1=CC=C(C(=O)N2CCC[C@@H](O)C3=CC(Cl)=CC=C23)C(C)=C1
IUPAC Name N-{4-[(5R)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1H-1-benzazepine-1-carbonyl]-3-methylphenyl}-2-methylbenzamide
Molecular Weight (g/mol) 448.95
Chemical Name or Material Tolvaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.