Learn More
Novus Biologicals™ TBC1D10A 293T Cell Transient Overexpression Lysate
Product Code. 18537181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18537181 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18537181 Supplier Novus Biologicals™ Supplier No. H00083874T01
 Restricted Item
Estimated Shipment: 16-10-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target TBC1D10A
Synonym EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10 A, TBC1 domain family, member 10 A
Gene Symbol TBC1D10A