Learn More
Tocris Bioscience™ Resiquimod
GREENER_CHOICE
Product Code. 18005712
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005712 10 mg 10mg
18055902 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18005712 Supplier Tocris Bioscience™ Supplier No. 4536/10
Toll-like receptor 7 (TLR7) agonist
Toll-like receptor 7 (TLR7) agonist. Induces upregulation of IL-6, IL-12, IFN-γ and iNOS expression in mouse bone marrow-derived macrophages (BMMs). Inhibits RANKL-induced osteoclast differentiation in mouse BMMs and human peripheral blood monocytes. Also promotes the differentiation of MDSCs into macrophages and dendritic cells. Proinflammatory and antiviral.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144875-48-9
Molecular Formula C17H22N4O2
Specifications
Quantity 10 mg
Formula Weight 314.38
Purity >98%
Molecular Weight (g/mol) 314.389
Chemical Name or Material Resiquimod