accessibility menu, dialog, popup

UserName

H3CONH2NNNH3CCH3HO

CAS RN 144875-48-9

IUPAC Name: 1-[4-amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol
Synonyms: resiquimod
Molecular Weight (g/mol): 314.39
Molecular Formula: C17H22N4O2
InChi Key: BXNMTOQRYBFHNZ-UHFFFAOYSA-N
SMILES: CCOCC1=NC2=C(N1CC(C)(C)O)C1=CC=CC=C1N=C2N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)