Learn More
Novus Biologicals™ Proteinase K
Product Code. 18700414 Shop All Bio Techne Products
Change view
Click to view available options
For Use With (Application):
Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
18700414 Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.) 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18700414 Supplier Novus Biologicals™ Supplier No. NB90066727
  • ONLINE EXCLUSIVE PRICE
    447.00 EUR
    422.10 EUR / 2mL
    Save 24.90 EUR
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
This is a 25X concentrated solution. For use, prepare a working solution: Place 100 uL Proteinase K solution in a glas tube, add 2.4 mL distilled water and mix. The optimal digestion conditions will vary depending on the fixation conditions and should be optimized by individual laboratory. However, incubation of tissue sections for 10-30 minutes at 37C is sufficient for in situ.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Sodium Azide is included at less than 0.1%. Exact amount not available.
For Use With (Application) Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)