Learn More
Novus Biologicals™ PAK4 Overexpression Lysate
Artikelnummer. 18256461 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Quantity:
0.1 mg
Packungsgröße:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
18256461 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18256461 Lieferant Novus Biologicals™ Lieferanten-Nr. NBL114081
 Restricted Item
Estimated Shipment: 08-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The PAK4 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC26847 and based on accession number NM_114831. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens p21 protein (Cdc42/Rac)-activated kinase 4 (PAK4), transcript variant 2, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native
For Research Use Only