Learn More
Melford ONPG
Product Code. 30018613
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Unit Size:
1g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018613 5 g 5g
30018611 1 g 1g
30018612 25 g 25g
30018614 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30018613 Supplier Melford Supplier No. N810005.0
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Specifications
Quantity 5 g