missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Oleoyl Coenzyme A Dipotassium Salt

Product Code. 11394099 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394099 1 mg 1mg
11304109 5 mg 5mg
11314109 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394099 Supplier MP Biomedicals™ Supplier No. 0210090401
  • ONLINE EXCLUSIVE PRICE
    66.15 EUR
    63.00 EUR / 1mg
    Save 3.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.

  • Coenzyme A functions as an acyl group carrier, acetyl-CoA

  • Coenzyme A functions as an acyl group carrier, acetyl-CoA
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage -20°C
Form Powder
Quantity 1 mg
Molecular Weight (g/mol) 1031.986
Product Type Biochemicals
Purity or Quality Grade ≥90%

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.