Learn More
Melford Methyl-B-D-Glucuronide Sodium Salt
Product Code. 30018494
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018494 Supplier Melford Supplier No. M638401.0
 Restricted Item
Estimated Shipment: 12-11-2026
Potential glucuronidase inducer.
Potential glucuronidase inducer. White fine crystalline powder.
Specifications
Quantity 1 g