Learn More
Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
Description
The Kidney Matched Tumor & Normal Tissue Lysate is derived from human.
Specifications
Specifications
| Tissue | Kidney |
| Quantity | 2 Vials |
| Host Species | Human |
| Content And Storage | Store at −80°C. Avoid freeze-thaw cycles. |