Learn More
Novus Biologicals™ EV Precipitation Solution (Urine)
Shop All Bio Techne Products
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
Specifications
Specifications
| Product Type | EV isolation, exosome isolation, extracellular vesicle isolation |
| Quantity | 30 mL |
| Sufficient For | A 30mL solution is equalivant to 25 reactions (urine) |
| Content And Storage | Store at room temperature. |
| Sample Type | Urine |