missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ D-Val-Leu-Lys-7-amino-4-methylcoumarin

Product Code. 11494686 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10mg
25mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494686 10mg 10mg
11404696 25mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494686 Supplier MP Biomedicals™ Supplier No. 03AMC08110
  • ONLINE EXCLUSIVE PRICE
    385.00 EUR
    366.45 EUR / 10mg
    Save 18.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

D-Val-Leu-Lys-7-amino-4-methylcoumarin

  • It is used for assay of tissue plasminogen activator.
  • The AMC fluorogenic substrates are widely used for routine assays of peptidases.
  • A short peptide sequence is conjugated to 7-AMINO-4-METHYLCOUMARIN, which is released upon cleavage by peptidase.
  • The release of 7-AMINO-4-METHYLCOUMARIN is then measured spectrophotometrically.
  • The D-VAL-LEU-LYS-7-AMINO-4-METHYLCOUMARIN is used as s substrate for Plasmin
TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) Apoptosis, Cell Viability Analysis, Cell Signalling
Quantity 10mg
Molecular Weight (g/mol) 515.4 (Not including 2TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Store at Room Temperature (15-30°C). Keep cool and dry.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.