Learn More
Tocris Bioscience™ Cyclopiazonic acid
GREENER_CHOICE
Product Code. 18772654
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772654 10 mg 10mg
18702724 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18772654 Supplier Tocris Bioscience™ Supplier No. 1235/10
  • ONLINE EXCLUSIVE PRICE
    222.00 EUR
    210.00 EUR / 10mg
    Save 12.00 EUR
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Inhibitor of SERCA ATPase
Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18172-33-3
Molecular Formula C20H20N2O3
Specifications
Quantity 10 mg
Formula Weight 336.39
Purity >98%
Molecular Weight (g/mol) 336.391
Chemical Name or Material Cyclopiazonic acid