CAS RN 18172-33-3
Cyclopiazonic acid, 98%, Thermo Scientific™
CAS: 18172-33-3 Molecular Formula: C20H20N2O3 Molecular Weight (g/mol): 336.391 MDL Number: MFCD00167445 InChI Key: SZINUGQCTHLQAZ-AFAVFJNCSA-N Synonym: cyclopiazonic acid,alpha-cyclopiazonic acid,unii-x9tly4580z,ccris 4942,alpha-cyclopiazonate,cyclopyazonic acid,6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione,9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha,6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one,ambotzls-1020 PubChem CID: 54711281 SMILES: CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
Cyclopiazonic Acid, >98%, MP Biomedicals
CAS: 18172-33-3 Molecular Formula: C20H20N2O3 Molecular Weight (g/mol): 336.391 InChI Key: SZINUGQCTHLQAZ-AFAVFJNCSA-N Synonym: cyclopiazonic acid,alpha-cyclopiazonic acid,unii-x9tly4580z,ccris 4942,alpha-cyclopiazonate,cyclopyazonic acid,6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione,9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha,6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one,ambotzls-1020 PubChem CID: 54711281 SMILES: CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
α-Cyclopiazonic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Cyclopiazonic Acid, TRC
CAS: 18172-33-3 Molecular Formula: C20 H20 N2 O3 Molecular Weight (g/mol): 336.38 Synonym: 9H-Pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, 10-acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-, (6aα,11aβ,11bα)- (8CI),(6aR,11aS,11bR)-10-Acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-9H-pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one,Cyclopiazonic acid,NSC 117181,α-Cyclopiazonic acid,α-Cyclopiazonic Acid,9H-Pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, 10-acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-, (6aα,11aβ,11bα)- (8CI),(6aR,11aS,11bR)-10-Acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-9H-pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one,Cyclopiazonic acid,NSC 117181,α-Cyclopiazonic acid SMILES: CC(=O)C1=C(O)[C@@H]2[C@@H]3[C@@H](Cc4cccc5[nH]cc3c45)C(C)(C)N2C1=O
Tocris Bioscience™ Cyclopiazonic acid
CAS: 18172-33-3 Molecular Formula: C20H20N2O3