missing translation for 'onlineSavingsMsg'
Learn More

Cyclopiazonic Acid, MP Biomedicals™

Product Code. 11494142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494142 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494142 Supplier MP Biomedicals Supplier No. 0215889510
  • ONLINE EXCLUSIVE PRICE
    239.00 EUR
    226.80 EUR / 10mg
    Save 12.20 EUR
    5% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Displays anti-arrhythmic effects. Cyclopiazonic Acid, MP Biomedicals inhibits the sarcoplasmic reticulum Ca2+-ATPase, which induces the release of intracellular stored Ca2+ without increasing IP3 levels.

Cyclopiazonic acid is a highly specific inhibitor of the Ca2+-ATPase of sarcoplasmic reticulum, completely inhibiting the enzyme at 6-8nmol/mg protein at the low concentration of 0.5-2μM. Cyclopiazonic acid on Langendorff-perfused murine hearts displays anti-arrhythmic effects. Also, administration on rat soleus muscle showed a decrease in hormone-sensitive lipase activity and triacylglycerol hydrolysis.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18172-33-3
Melting Point 237°C to 238°C (literature)
Molecular Formula C20H20N2O3
Quantity 10 mg
Synonym cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
Solubility Information Soluble in chloroform (10mg/mL) or methanol (10mg/mL, clear, bright yellow solution).
InChI Key SZINUGQCTHLQAZ-AFAVFJNCSA-N
SMILES CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
Molecular Weight (g/mol) 336.391
PubChem CID 54711281
Formula Weight 336.4
Percent Purity >98%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.