Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Product Code. 18113923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18113923 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18113923 Supplier Novus Biologicals™ Supplier No. NBP231142SS
  • ONLINE EXCLUSIVE PRICE
    194.00 EUR
    182.70 EUR / 25µg
    Save 11.30 EUR
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Specifications
Concentration 1.0 mg/mL
Content And Storage Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Quantity 0.025 mg
For Use With (Application) Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 μg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.)
Target CpG Oligodeoxynucleotides Mouse
Type CpG Oligodeoxynucleotides