Learn More
Novus Biologicals™ Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Product Code. 18197489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Units
Unit Size:
2 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18197489 2 Units 2 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18197489 Supplier Novus Biologicals™ Supplier No. NBL126503
  • ONLINE EXCLUSIVE PRICE
    481.00 EUR
    454.65 EUR / Pack of 2
    Save 26.35 EUR
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
The Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from chicken.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 Units
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Chicken Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Chicken
Host Species Chicken
Product Type Chicken Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Tissue Universal
Show More Show Less
For Research Use Only