missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, 98+%, Thermo Scientific™

131.00 EUR - 230.00 EUR

Chemical Identifiers

CAS 603-50-9
Molecular Formula C22H19NO4
Molecular Weight (g/mol) 361.40
MDL Number MFCD00038039
InChI Key KHOITXIGCFIULA-UHFFFAOYSA-N
Synonym bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
PubChem CID 2391
IUPAC Name 4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444469
SDS View Documents
Thermo Scientific Alfa Aesar
J61997.09
10g
131.00 EUR
10g
View alternative products
Save Up to 
»
Estimated Shipment: 31-08-2026
Add to basket
 
 Restricted Item
15454469
SDS View Documents
Thermo Scientific Alfa Aesar
J61997.14
25g
230.00 EUR
25g
View alternative products
Save Up to 
»
Estimated Shipment: 31-08-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in alcohol (slightly), ether (slightly), chloroform, and acetone. Insoluble in water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
Specifications

Chemical Identifiers

603-50-9
361.40
KHOITXIGCFIULA-UHFFFAOYSA-N
2391
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
C22H19NO4
MFCD00038039
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate

Specifications

603-50-9
MFCD00038039
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
KHOITXIGCFIULA-UHFFFAOYSA-N
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
2391
≥98%
C22H19NO4
14,1242
Soluble in alcohol (slightly),ether (slightly),chloroform,and acetone. Insoluble in water.
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1
361.40
361.4
Bisacodyl
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 210-044-4

missing translation for 'rtecsNumber' : SM8750000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.